Learn More
5-Hydroxypicolinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxypicolinic Acid |
| CAS | 15069-92-8 |
| Molecular Formula | C6 H5 N O3 |
| Quantity | 25 g |
| InChI Formula | InChI=1S/C6H5NO3/c8-4-1-2-5(6(9)10)7-3-4/h1-3,8H,(H,9,10) |
| SMILES | OC(=O)c1ccc(O)cn1 |
| IUPAC Name | 5-hydroxypyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 139.11 |
| Formula Weight | 139.0269 |