Learn More
5-Hydroxyadamantan-2-one, 98%, BLD Pharm™
Product Code. 31073321
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31073321 1 g
31072858 100 g
31058217 500 g
31058023 10 g
31059134 5 g
31065726 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31073321 Supplier BLD Pharm Supplier No. BD1041931g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 20098-14-0
Molecular Formula C10H14O2
MDL Number MFCD00192211
Quantity 1 g
SMILES O=C1C2CC3CC(C2)(O)CC1C3
Molecular Weight (g/mol) 166.22
Percent Purity 98%
Chemical Name or Material 5-Hydroxyadamantan-2-one