missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hmC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-59184
This item is not returnable.
View return policy
Description
5-hmC Polyclonal specifically detects 5-hmC in Human, Mouse, Xenopus, Zebrafish samples. It is validated for ELISA, Dot Blot, Methylated DNA Immunoprecipitation.
Specifications
| 5-hmC | |
| Polyclonal | |
| ELISA 1:100 - 1:500, Dot Blot 1:1000, Methylated DNA Immunoprecipitation 3.5 μg/IP | |
| 5'-hmC, 5-hydroxymethylcytosine | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Xenopus species, Zebrafish | |
| Purified |
| ChIP Assay, Dot Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
| HMCES | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| DNA Repair, Epigenetics, Stem Cells | |
| 56941 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction