Learn More
5-Hexynoic acid, 96%
Product Code. 11406187
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11406187 1 g 1g
11416187 5 g 5g
11426187 25 g 25g
3 options
Product Code. 11406187
Brand: Thermo Scientific Alfa Aesar B25132.03
Request Bulk or Custom Format
CAS: 53293-00-8 | C6H8O2 | 112.128 g/mol
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O
Specifications
Density 0.99
Boiling Point 73°C to 74°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4495
Quantity 1 g
UN Number UN3265
Beilstein 1743192
Solubility Information Miscible with water.
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 5-Hexynoic acid