Learn More
5-Fluoro-m-xylene, 97%
Product Code. 11457267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11457267 1 g 1g
11467267 5 g 5g
2 options
Product Code. 11457267
Brand: Thermo Scientific Alfa Aesar A15020.03
Request Bulk or Custom Format
CAS: 461-97-2 | C8H9F | 124.158 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 461-97-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00052366
InChI Key RCWIWNUVHNAUQC-UHFFFAOYSA-N
Synonym 3,5-dimethylfluorobenzene, 5-fluoro-m-xylene, 1,3-dimethyl-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethyl, pubchem4410, 5-fluoro-1,3-xylene, acmc-1ahss, 3,5-dimethyl-fluorobenzene, 3,5-dimethylphenyl fluoride, ksc497s5r
PubChem CID 521192
IUPAC Name 1-fluoro-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)F)C
Specifications
Boiling Point 144°C to 145°C
Quantity 1 g
UN Number UN1993
Beilstein 1926374
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 5-Fluoro-m-xylene