Learn More
5-Fluoro-2-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
Product Code. 15475157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475157 1 g 1g
15485157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15475157 Supplier Thermo Scientific Alfa Aesar Supplier No. H32445.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 243459-91-8 | Molecular Formula: C7H4F4O | Molecular Weight: 180.102 g/mol
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
Solubility
Sparingly Soluble (0.40 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 243459-91-8
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00236302
InChI Key QWLZSSYHAJSEHU-UHFFFAOYSA-N
PubChem CID 2774803
IUPAC Name 5-fluoro-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1F)O)C(F)(F)F
Specifications
Melting Point 48°C to 52°C
Quantity 1 g
UN Number UN3261
Synonym 5-fluoro-2-trifluoromethyl phenol, 5-fluoro-2-trifluoromethyl-phenol, 4-fluoro-2-hydroxybenzotrifluoride, pubchem2171, intermediates-zcf02657, phenol,5-fluoro-2-trifluoromethyl, phenol, 5-fluoro-2-trifluoromethyl
Solubility Information Sparingly Soluble (0.40g/L) (25°C).
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)phenol