Learn More
Invitrogen™ 5' Fluorescent Labeled Oligo
Product Code. 15521134
Change view
Click to view available options
Product Type:
5' Fluorescent Labeled Single Primer
5' Labeled Primers
Custom 5' Fluorescent Labeled Oligo Primers
Custom Prism Primer
Oligonucleotides, Primers
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
1 set
300pmol
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
15521134 5' Fluorescent Labeled Single Primer 300,000 pmol 300pmol
11563827 Oligonucleotides, Primers 10,000 pmol Each
15438394 5' Labeled Primers 80,000 pmol 1 set
11573837 Custom 5' Fluorescent Labeled Oligo Primers 80,000 pmol Each
11539446 Custom Prism Primer 300,000 pmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15521134 Supplier Invitrogen™ Supplier No. 450017H
 Restricted Item
Estimated Shipment: 05-11-2026
5' Fluorescent Labeled Oligo
Custom 5'-labeled primers are ideal for applications including fragment analysis, gene expression studies, genotyping, and PCR. Choose among FAM™, TET™, VIC™, HEX™, NED™, and PET™ as fluorescent 5' labels. Oligos are desalted; HPLC purification is also available.
TRUSTED_SUSTAINABILITY
Specifications
Description 5' Fluorescent Labeled Single Primer, 300 kpmol
Product Type 5' Fluorescent Labeled Single Primer
Length (Metric) 1.524 m
Content And Storage Store in Freezer (-5°C to -30°C).
Purification Method Desalted, HPLC
Label or Dye FAM, HEX, NED, PET, TET, VIC
Primer 5' Labeled
Quantity 300,000 pmol
Grade HPLC
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR