Learn More
5-Ethyl-2-furaldehyde, 98%, Thermo Scientific Chemicals
Catalog Number 11356946
Click to view available options
Quantity:
1 g
5 g
23074-10-4
C7H8O2
124.14
MFCD00030605
XADGZBXFWQHBDB-UHFFFAOYSA-N
89989
5-ethylfuran-2-carbaldehyde
CCC1=CC=C(O1)C=O
23074-10-4
C7H8O2
124.14
MFCD00030605
XADGZBXFWQHBDB-UHFFFAOYSA-N
89989
5-ethylfuran-2-carbaldehyde
CCC1=CC=C(O1)C=O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.055
Boiling Point 79°C to 81°C (12 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.522
Quantity 1 g
Beilstein 1982
Sensitivity Air Sensitive
Synonym 5-ethyl-2-furaldehyde, 5-ethylfurfural, 2-furancarboxaldehyde, 5-ethyl, 5-ethyl-furan-2-carbaldehyde, 2-furancarboxaldehyde,5-ethyl, 5-ethylfurfuraldehyde, acmc-1cc1a, 5-ethyl-2-furancarboxaldehyde, 5-ethylfuran-2-carboxaldehyde
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 5-Ethyl-2-furaldehyde