missing translation for 'onlineSavingsMsg'
Learn More

5-(Difluoromethoxy)-3-fluorobenzyl bromide, JRD, 97%, Thermo Scientific™

Catalog Number p-7031782
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017779-39-3
C8H6BrF3O
255.034
MFCD09832362
SZFFWIBFEJPELW-UHFFFAOYSA-N
1-bromomethyl-3-difluoromethoxy-5-fluorobenzene, 5-difluoromethoxy-3-fluorobenzyl bromide, jrd, 5-difluoromethoxy-3-fluorobenzyl bromide, 5-bromomethyl-3-fluorophenoxy difluoromethane
46737678
1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene
C1=C(C=C(C=C1OC(F)F)F)CBr
This item is not returnable. View return policy
1017779-39-3
C8H6BrF3O
255.034
MFCD09832362
SZFFWIBFEJPELW-UHFFFAOYSA-N
1-bromomethyl-3-difluoromethoxy-5-fluorobenzene, 5-difluoromethoxy-3-fluorobenzyl bromide, jrd, 5-difluoromethoxy-3-fluorobenzyl bromide, 5-bromomethyl-3-fluorophenoxy difluoromethane
46737678
1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene
C1=C(C=C(C=C1OC(F)F)F)CBr

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 5-(Difluoromethoxy)-3-fluorobenzyl bromide, JRD
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'