missing translation for 'onlineSavingsMsg'
Learn More

5-Di-tert-butylphosphino-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 97%

Product Code. p-7055318
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15488556

Brand: Thermo Scientific Alfa Aesar H63864.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 894086-00-1 | C32H35N4P | 506.634 g/mol

5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Di-tert-butylphosphino)-1′, 3′, 5′-triphenyl-1′H-[1,4′]bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 894086-00-1
Molecular Formula C32H35N4P
Molecular Weight (g/mol) 506.634
MDL Number MFCD09038440
InChI Key PTXJGGGNGMPMBG-UHFFFAOYSA-N
Synonym bippyphos, t-bu-bippyphos, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4'-bipyrazole, 5-di-tert-butylphosphino-1', 3', 5'-triphenyl-1'h-1,4' bipyrazole, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4' bipyrazole, 5-di-t-butylphosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-bis tert-butyl phosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-di-tert-butylphosphanyl-1',3',5'-triphenyl-1,4'-bipyrazole, 1',3',5'-triphenyl-5-ditert-butylphosphino-1,4'-bi 1h-pyrazole, 5-di-tert-butyl-phosphanyl-1',3',5'-triphenyl-1'h-1,4' bipyrazolyl
PubChem CID 11547931
IUPAC Name ditert-butyl-[2-(1,3,5-triphenylpyrazol-4-yl)pyrazol-3-yl]phosphane
SMILES CC(C)(C)P(C1=CC=NN1C2=C(N(N=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C(C)(C)C

Specifications

Melting Point 169°C to 173°C
Color White
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 506.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Di-tert-butylphosphino-1',3',5'-triphenyl-1'H-1,4'-bipyrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.