Learn More
5-Decyne, 98%
Product Code. p-7020928
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11429993
Brand: Thermo Scientific Alfa Aesar A13050.09
Request Bulk or Custom Format
CAS: 1942-46-7 | C10H18 | 138.254 g/mol
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1942-46-7
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00027277
InChI Key JWBQJUFCNOLNNC-UHFFFAOYSA-N
Synonym 5-decyne, dibutylacetylene, 1,2-dibutylacetylene, acmc-209ey1, 5-decyne 8ci 9ci, 3,3?-diheptyloxacarbocyanine iodide
PubChem CID 16030
IUPAC Name dec-5-yne
SMILES CCCCC#CCCCC
Specifications
Density 0.77
Boiling Point 177°C to 178°C
Flash Point 54°C (129°F)
Odor Petrol-like
Refractive Index 1.433
Quantity 10 g
UN Number UN3295
Beilstein 1697840
Solubility Information Not miscible in water.
Formula Weight 138.26
Percent Purity 98%
Chemical Name or Material 5-Decyne