Learn More
5-Cyclopropylisoxazole-3-carboxylic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 110256-15-0 |
| Molecular Formula | C7H7NO3 |
| MDL Number | MFCD05668325 |
| Quantity | 1 g |
| SMILES | O=C(C1=NOC(C2CC2)=C1)O |
| Molecular Weight (g/mol) | 153.14 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Cyclopropylisoxazole-3-carboxylic acid |