Learn More
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid, 94%, Thermo Scientific™
Product Code. p-7033237
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11479460 250 mg 250mg
11489460 1 g 1g
2 options
Product Code. 11479460
Brand: Thermo Scientific Alfa Aesar H50306.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid is used as organic chemical synthesis intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 401629-04-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD04967194
InChI Key GNWMHLBUCXEXPE-UHFFFAOYSA-N
Synonym 3-cyclopropyl-1h-pyrazole-5-carboxylic acid, 3-cyclopropylpyrazole-5-carboxylic acid, 5-cyclopropyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-cyclopropyl, 5-cyclopropylpyrazole-3-carboxylic acid, acmc-1ap8l, 1h-pyrazole-3-carboxylic acid, 5-cyclopropyl, 1h-pyrazole-3-carboxylic acid, 5-cyclopropyl-9ci
PubChem CID 776464
IUPAC Name 5-cyclopropyl-1H-pyrazole-3-carboxylic acid
SMILES C1CC1C2=CC(=NN2)C(=O)O
Specifications
Melting Point ∽175°C (decomposition)
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 94%
Chemical Name or Material 5-Cyclopropyl-1H-pyrazole-3-carboxylic acid