Learn More
5-Cyanopyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Catalog Number 15496567
Click to view available options
Quantity:
250 mg
1 g
53234-55-2
C7H4N2O2
148.12
MFCD09152775
HRLVPHGCEGTVLK-UHFFFAOYSA-N
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
12400734
5-cyanopyridine-2-carboxylic acid
OC(=O)C1=NC=C(C=C1)C#N
53234-55-2
C7H4N2O2
148.12
MFCD09152775
HRLVPHGCEGTVLK-UHFFFAOYSA-N
5-cyanopicolinic acid, 5-cyano-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-cyano, 5-cyano-2-pyridincarboxylic acid, 5-cyanopicolinicacid, 5-cyano-picolinic acid, pubchem16740, acmc-1cubt, 5-cyano-2-picolinic acid, 5-cyano-2-pyridine carboxylic acid
12400734
5-cyanopyridine-2-carboxylic acid
OC(=O)C1=NC=C(C=C1)C#N
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 212°C to 214°C
Quantity 250 mg
UN Number UN3439
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 148.1
Percent Purity 95%
Chemical Name or Material 5-Cyanopyridine-2-carboxylic acid