missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosulfonyl-2-methoxybenzoic acid, 99%, Thermo Scientific™

Product Code. 11335359
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335359 1 g 1g
11345359 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335359 Supplier Thermo Scientific Alfa Aesar Supplier No. H50376.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51904-91-7
Molecular Formula C8H7ClO5S
Molecular Weight (g/mol) 250.649
MDL Number MFCD00066912
InChI Key OUBZCDOQEMLMAB-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-methoxybenzoic acid, 5-chlorosulphonyl-2-anisic acid, benzoic acid, 5-chlorosulfonyl-2-methoxy, 5-chlorosulphonyl-2-methoxybenzoic acid, 5-chlorosulfonyl-2-methoxybenzoicacid, acmc-1ar14, 2-methoxy-5-chlorosulfonylbenzoic acid, 5-chlorosulfonyl-2-methoxy-benzoic acid, 5-chlorosulphonyl-2-methoxy benzoic acid, 5-chlorosulphonyl-2-methoxy-benzoic acid
PubChem CID 104007
IUPAC Name 5-chlorosulfonyl-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(=O)O

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 250.66
Percent Purity 99%
Chemical Name or Material 5-Chlorosulfonyl-2-methoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.