Learn More
5-Chloro-2-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
Product Code. 15434177
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15434177 5 g 5g
15424177 1 g 1g
2 options
Product Code. 15434177
Brand: Thermo Scientific Alfa Aesar H31584.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1092461-15-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD11519353
InChI Key VJRCCSALVWMZAN-UHFFFAOYSA-N
Synonym 5-chloro-2-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-5-chlorobenzaldehyde, 5-chloro-2-trifluoromethoxy benzaldehyde, jrd
PubChem CID 46835525
IUPAC Name 5-chloro-2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=C(C=O)C=C(Cl)C=C1
Specifications
Refractive Index 1.4755
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-(trifluoromethoxy)benzaldehyde