Learn More
5-Chloro-1-pentanol, TRC
Product Code. 30368499
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368499 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30368499 Supplier Toronto Research Chemicals Supplier No. TRCC35034010G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Chloro-1-pentanol
CAS 5259-98-3
Molecular Formula C5H11ClO
Quantity 10 g
Synonym 5-Chloro-1-pentanol, 1-Chloro-5-pentanol, 5-Chloropentanol, NSC 1107, Pentamethylene Chlorohydrin, ω-Chloropentanol
InChI Formula InChI=1S/C5H11ClO/c6-4-2-1-3-5-7/h7H,1-5H2
SMILES OCCCCCCl
IUPAC Name 5-chloropentan-1-ol
Molecular Weight (g/mol) 122.59
Formula Weight 122.0498