missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Glucuronide Cyclohexylammonium Salt, MP Biomedicals™

Catalog Number 11448262
Click to view available options
Quantity:
5 mg
10 mg
25 mg
This item is not returnable. View return policy
18656-96-7
C20H26BrClN2O7
521.789
JXCKZXHCJOVIAV-CYRSAHDMSA-N
c14h13brclno7.c6h13n, cyclohexanaminium 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexylammonium salt, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexyl ammonium salt, 5-bromo-4-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt monohydrate
42609655
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium
C1CCC(CC1)[NH3+].C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br
This item is not returnable. View return policy
18656-96-7
C20H26BrClN2O7
521.789
JXCKZXHCJOVIAV-CYRSAHDMSA-N
c14h13brclno7.c6h13n, cyclohexanaminium 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexylammonium salt, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexyl ammonium salt, 5-bromo-4-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt monohydrate
42609655
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium
C1CCC(CC1)[NH3+].C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br

Product is a β-glucuronidase substrate, which forms an intense blue precipitate upon enzymatic action. Product is used for the detection of the GUS gene in bacterial colonies and in histochemical applications.

Melting Point 241°C to 243°C (literature)
Quantity 5 mg
Solubility Information Soluble in DMF (2mg/mL; clear, colorless solution).
Optical Rotation (c=1, DMF : H2O 1:1): –90.4 db (Lit.)
Formula Weight 521.8g/mol
Percent Purity ≥95%
Physical Form Crystalline Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'