missing translation for 'onlineSavingsMsg'
Learn More

X-Gal, 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, Fisher BioReagents™

Product Code. p-24858
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass Bottle
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10234923 100 mg Glass Bottle 100mg
10113253 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234923 Supplier Fisher BioReagents Supplier No. BP1615100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For blue/white colony screening, immunoblotting and immunocytochemical assays

  • Chromogenic substrate used for blue-white selection of recombinant bacterial colonies with the lac+ genotype
  • Forms blue precipitate when hydrolyzed by β-galactosidase
  • Soluble in DMF and Methanol
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Especificaciones

Alpha Vector Bromo-4-chloro-3-indolyl-[beta]-D-galactopyranoside (X-Gal)
Melting Point 230°C
Color White
Packaging Glass Bottle
Quantity 100 mg
Merck Index 15, 10272
Specific Rotation -56 to -64° (+ or -)
Formula Weight 408.63
Physical Form Powder or Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl- β-D-galactopyranoside
compliance-icons
Nombre del producto
  • 5-Bromo-4-chloro-3-indolyl-b-D-galactopyranoside
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.