missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-idolyl-α-D-galactopyranoside, MP Biomedicals™

Product Code. 11496331 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496331 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496331 Supplier MP Biomedicals Supplier No. 0219392125
  • ONLINE EXCLUSIVE PRICE
    963.00 NOK
    916.50 NOK / 25mg
    Save 46.50 NOK
    4% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity substrate for research applications.

Specifications

CAS 107021-38-5
Color White
Assay Percent Range >99%
Molecular Formula C14H15BrClNO6
MDL Number MFCD00063780
Quantity 25 mg
Synonym x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside
InChI Key OPIFSICVWOWJMJ-UHFFFAOYNA-N
SMILES OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O
IUPAC Name 2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 408.63
PubChem CID 10173103
ChEBI CHEBI:75506
Formula Weight 408.63
Grade Research, NA
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-idolyl-α-D-galactopyranoside
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.