missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine, 95%, Thermo Scientific™

Catalog Number 15427695
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
245765-92-8
C12H17BrN2O
285.185
MFCD09037448
MKBZSCNCWLNWFH-UHFFFAOYSA-N
5-bromo-3-methyl-2-n,2,2,2-tetramethylacetamido pyridine, n-5-bromo-3-methylpyridin-2-yl-n,2,2-trimethylpropanamide, n-5-bromo-3-methylpyridin-2-yl-n-methylpivalamide, n-5-bromo-3-methylpyridin-2-yl-n-methyl-2,2-dimethylpropanamide, 5-bromo-3-methyl-2-methyl-pivaloylamino pyridine, acmc-20akaw, propanamide,n-5-bromo-3-methyl-2-pyridinyl-n,2,2-trimethyl
18329348
N-(5-bromo-3-methylpyridin-2-yl)-N,2,2-trimethylpropanamide
CC1=CC(=CN=C1N(C)C(=O)C(C)(C)C)Br
This item is not returnable. View return policy
245765-92-8
C12H17BrN2O
285.185
MFCD09037448
MKBZSCNCWLNWFH-UHFFFAOYSA-N
5-bromo-3-methyl-2-n,2,2,2-tetramethylacetamido pyridine, n-5-bromo-3-methylpyridin-2-yl-n,2,2-trimethylpropanamide, n-5-bromo-3-methylpyridin-2-yl-n-methylpivalamide, n-5-bromo-3-methylpyridin-2-yl-n-methyl-2,2-dimethylpropanamide, 5-bromo-3-methyl-2-methyl-pivaloylamino pyridine, acmc-20akaw, propanamide,n-5-bromo-3-methyl-2-pyridinyl-n,2,2-trimethyl
18329348
N-(5-bromo-3-methylpyridin-2-yl)-N,2,2-trimethylpropanamide
CC1=CC(=CN=C1N(C)C(=O)C(C)(C)C)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 285.18
Percent Purity 95%
Chemical Name or Material 5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'