Learn More
5-Bromo-3-indolyl beta-d-galactopyranoside, 98+%
Product Code. 15427169
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. 15427169
Brand: Thermo Scientific Alfa Aesar J65978.03
Request Bulk or Custom Format
alpha-Galactosidase substrate for Lac-gene detection systems | CAS: 97753-82-7 | C14H16BrNO6 | 374.19 g/mol
5-Bromo-3-indolyl-β-D-galactopyranoside is an α-galactosidase substrate which is converted to an insoluble indigo-blue chromophore darker than that released by X-GAL. It is ideal for Lac gene detection systems in immunoblotting, immunocytochemical, and histological applications. Also used as chromogenic substrate that is suitable for identification of lac-(+) bacterial colonies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-3-indolyl-β-D-galactopyranoside is an α-galactosidase substrate which is converted to an insoluble indigo-blue chromophore darker than that released by X-GAL. It is ideal for Lac gene detection systems in immunoblotting, immunocytochemical, and histological applications. Also used as chromogenic substrate that is suitable for identification of lac-(+) bacterial colonies.
Solubility
Soluble in dimethylformamide (100 mg/ml), dimethylformamide/water (1:1 v/v) (10 mg/ml - clear, practically colorless solution).
Notes
Store in freezer (-20°C). Keep the container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97753-82-7
Molecular Formula C14H16BrNO6
Molecular Weight (g/mol) 374.19
MDL Number MFCD00063691
InChI Key LINMATFDVHBYOS-MBJXGIAVSA-N
Synonym 5-bromo-3-indolyl-beta-d-galactopyranoside, 5-bromo-3-indolyl-b-d-galactopyranoside, bluo-gal, blue-gal, 2s,3r,4s,5r,6r-2-5-bromo-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 5-brig, blue gal, 5-bromo-3-indolyl-beta-galactoside, 5-bromo-3-indolyl beta-d-galactopyranoside, 5-bromo-3-indoxyl-beta-d-galactopyranoside
PubChem CID 126898
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O
Specifications
Color White
Quantity 1 g
Solubility Information Soluble in dimethylformamide (100mg/ml),dimethylformamide/water (1:1 v/v) (10mg/ml- clear,practically colorless solution).
Formula Weight 374.18
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 5-Bromo-3-indolyl beta-d-galactopyranoside