missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15494367
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
923281-52-1
C8H4BrF3O2
269.02
MFCD03701029
IBEWLWBZTHFCPI-UHFFFAOYSA-N
5-bromo-2-trifluoromethoxy benzaldehyde, benzaldehyde, 5-bromo-2-trifluoromethoxy, acmc-209rfd, 2-trifluoromethoxy-5-bromobenzaldehyde, 4-bromo-2-formyl-alpha,alpha,alpha-trifluoroanisole
26599201
5-bromo-2-(trifluoromethoxy)benzaldehyde
FC(F)(F)OC1=C(C=O)C=C(Br)C=C1
This item is not returnable. View return policy
923281-52-1
C8H4BrF3O2
269.02
MFCD03701029
IBEWLWBZTHFCPI-UHFFFAOYSA-N
5-bromo-2-trifluoromethoxy benzaldehyde, benzaldehyde, 5-bromo-2-trifluoromethoxy, acmc-209rfd, 2-trifluoromethoxy-5-bromobenzaldehyde, 4-bromo-2-formyl-alpha,alpha,alpha-trifluoroanisole
26599201
5-bromo-2-(trifluoromethoxy)benzaldehyde
FC(F)(F)OC1=C(C=O)C=C(Br)C=C1

5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 269.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'