Learn More
5-Bromo-2-fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific™
Product Code. 15446286
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446286 250 mg 250mg
15447628 1 g 1g
15457628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15446286 Supplier Thermo Scientific Alfa Aesar Supplier No. H61123.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. Afficher la politique du retour.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 875781-15-0
Molecular Formula C6H3BrFNO
Molecular Weight (g/mol) 203.998
MDL Number MFCD09842440
InChI Key MUYVOGAJRCBWCY-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoronicotinaldehyde, 5-bromo-2-fluoropyridine-3-carboxaldehyde, 5-bromo-2-fluoro-3-pyridinecarboxaldehyde, 5-bromo-2-fluoro-pyridine-3-carbaldehyde, 5-bromo-2-fluoro-3-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo-2-fluoro, pubchem17162, acmc-209qoh, ksc495s8t
PubChem CID 45480192
IUPAC Name 5-bromo-2-fluoropyridine-3-carbaldehyde
SMILES C1=C(C(=NC=C1Br)F)C=O
Spécification
Color White to Yellow
Quantity 250 mg
Beilstein 14515665
Formula Weight 204
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-fluoropyridine-3-carboxaldehyde
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.