missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxypyridine-3-boronic acid, 97%, Thermo Scientific™

Catalog Number 15420058
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871332-98-8
C7H9BBrNO3
245.87
MFCD07363847
VXXXKWUXSALQQW-UHFFFAOYSA-N
5-bromo-2-ethoxypyridine-3-boronic acid, 5-bromo-2-ethoxypyridin-3-yl boronic acid, 5-bromo-2-ethoxy-3-pyridyl boronic acid, 5-bromo-2-ethoxy-3-pyridineboronic acid, boronic acid,b-5-bromo-2-ethoxy-3-pyridinyl, acmc-209qhr, 5-bromo-2-ethoxypyridin-3-ylboronicacid, abbypharma ap-12-5495, 5-bromo-2-ethoxy-3-pyridinyl boronic acid, 5-bromanyl-2-ethoxy-pyridin-3-yl boronic acid
44886948
(5-bromo-2-ethoxypyridin-3-yl)boronic acid
CCOC1=NC=C(Br)C=C1B(O)O
This item is not returnable. View return policy
871332-98-8
C7H9BBrNO3
245.87
MFCD07363847
VXXXKWUXSALQQW-UHFFFAOYSA-N
5-bromo-2-ethoxypyridine-3-boronic acid, 5-bromo-2-ethoxypyridin-3-yl boronic acid, 5-bromo-2-ethoxy-3-pyridyl boronic acid, 5-bromo-2-ethoxy-3-pyridineboronic acid, boronic acid,b-5-bromo-2-ethoxy-3-pyridinyl, acmc-209qhr, 5-bromo-2-ethoxypyridin-3-ylboronicacid, abbypharma ap-12-5495, 5-bromo-2-ethoxy-3-pyridinyl boronic acid, 5-bromanyl-2-ethoxy-pyridin-3-yl boronic acid
44886948
(5-bromo-2-ethoxypyridin-3-yl)boronic acid
CCOC1=NC=C(Br)C=C1B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 152°C to 158°C
Quantity 250 mg
Formula Weight 245.87
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-ethoxypyridine-3-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'