Learn More
5-Bromo-2,4-dimethoxybenzoic acid, 97%
Product Code. p-7040744
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15457078 1 g 1g
15467078 5 g 5g
2 options
Product Code. 15457078
Brand: Thermo Scientific Alfa Aesar H60599.03
Request Bulk or Custom Format
CAS: 32246-20-1 | C9H9BrO4 | 261.071 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32246-20-1
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD05865172
InChI Key WOPJFSQYDOHZMK-UHFFFAOYSA-N
Synonym 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid
PubChem CID 13505097
IUPAC Name 5-bromo-2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Br)OC
Specifications
Melting Point 198°C to 202°C
Quantity 1 g
Formula Weight 261.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,4-dimethoxybenzoic acid