missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(2-thienyl)pyridine, 95%, Thermo Scientific™

Catalog Number 15403647
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
91891-74-6
C9H6BrNS
240.12
MFCD09037419
POYKHSYCYNRTKC-UHFFFAOYSA-N
5-bromo-2-2-thienyl pyridine, 5-bromo-2-thiophen-2-yl pyridine, 5-bromo-2-thien-2-yl pyridine, 2-2-thienyl-5-bromopyridine, 2-5-bromo-2-pyridyl thiophene
13238694
BrC1=CN=C(C=C1)C1=CC=CS1
This item is not returnable. View return policy
91891-74-6
C9H6BrNS
240.12
MFCD09037419
POYKHSYCYNRTKC-UHFFFAOYSA-N
5-bromo-2-2-thienyl pyridine, 5-bromo-2-thiophen-2-yl pyridine, 5-bromo-2-thien-2-yl pyridine, 2-2-thienyl-5-bromopyridine, 2-5-bromo-2-pyridyl thiophene
13238694
BrC1=CN=C(C=C1)C1=CC=CS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
IUPAC Name 5-bromo-2-thiophen-2-ylpyridine
Formula Weight 240.12
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(2-thienyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'