Learn More
5-Aminovaleric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Aminovaleric Acid |
| CAS | 660-88-8 |
| Molecular Formula | C5H11NO2 |
| Quantity | 5 g |
| Synonym | 5-Aminopentanoic Acid, 5-Amino-n-valeric Acid, Aminovaleric Acid, DAVA, NSC 58383, NSC 73123, delta-Amino-n-valeric Acid, delta-Aminopentanoic Acid, delta-Aminovaleric Acid, ω-Aminopentanoic Acid, ω-Aminovaleric Acid |
| InChI Formula | InChI=1S/C5H11NO2/c6-4-2-1-3-5(7)8/h1-4,6H2,(H,7,8) |
| SMILES | NCCCCC(=O)O |
| IUPAC Name | 5-aminopentanoic acid |
| Molecular Weight (g/mol) | 117.15 |
| Formula Weight | 117.079 |