Learn More
5-Aminopyrazole-1-carboxylic Acid Tert-butyl Ester O, TRC
Product Code. 30323587
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323587 Supplier Toronto Research Chemicals Supplier No. TRCA6291485MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Aminopyrazole-1-carboxylic Acid Tert-butyl Ester O
CAS 1003011-02-6
Molecular Formula C8H13N3O2
Quantity 5 mg
Synonym 1,1-Dimethylethyl Ester-5-amino-1H-pyrazole-1-carboxylic Acid
InChI Formula InChI=1S/C8H13N3O2/c1-8(2,3)13-7(12)11-6(9)4-5-10-11/h4-5H,9H2,1-3H3
SMILES CC(C)(C)OC(=O)n1nccc1N
IUPAC Name tert-butyl 5-aminopyrazole-1-carboxylate
Molecular Weight (g/mol) 183.21
Formula Weight 183.1008