Learn More
5-Aminonaphthalen-2-ol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 86-97-5 |
| Molecular Formula | C10H9NO |
| MDL Number | MFCD00035729 |
| Quantity | 25 g |
| SMILES | OC1=CC=C2C(N)=CC=CC2=C1 |
| Molecular Weight (g/mol) | 159.18 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Aminonaphthalen-2-ol |