Learn More
5-Acetyl-2,3-dihydrobenzo[b]furan, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 90843-31-5 |
| Molecular Formula | C10H10O2 |
| MDL Number | MFCD00068033 |
| Quantity | 25 g |
| SMILES | O=C(C1=CC=C2OCCC2=C1)C |
| Molecular Weight (g/mol) | 162.19 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Acetyl-2,3-dihydrobenzo[b]furan |