Learn More
5,7-Dibromoquinolin-8-ol, TRC
Product Code. 30377463
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377463 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30377463 Supplier Toronto Research Chemicals Supplier No. TRCD426143100MG
 Restricted Item
Estimated Shipment: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Broxyquinoline
CAS 521-74-4
Molecular Formula C9 H5 Br2 N O
Quantity 100 mg
Synonym 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol, 5,7-Dibromooxine, Brodiar, Broxyquinolin, Broxyquinoline, Colepur, Colipar, Dibromoksin, Dibromoquin, Dibromoxin, Dibromoxine, Dibromoxyquinoline, Fenilor, Intensopan, NSC 1810, NSC 74937, Paramiba, Paramibe, 8-Quinolinol, 5,7-dibromo-
InChI Formula InChI=1S/C9H5Br2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
SMILES Oc1c(Br)cc(Br)c2cccnc12
IUPAC Name 5,7-dibromoquinolin-8-ol
Molecular Weight (g/mol) 302.95
Formula Weight 300.8738