Learn More
5,7,3',4'-Tetra-O-benzylepicatechin-4Beta→8-5,7,3',4'-tetra-O-benzylepicatechin, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)-[4,8'-bi-2H-1-benzopyran]-3,3'-diol |
| CAS | 223387-28-8 |
| Molecular Formula | C8 H6 O3 . H2 O |
| Quantity | 0.5 mg |
| Synonym | (2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)-[4,8'-bi-2H-1-benzopyran]-3,3'-diol, 5,7,3',4'-Tetra-O-benzyl-(-)-epicatechin-(4β,8)-[5,7,3',4'-tetra-O-benzyl-(-)-epicatechin], 5,7,3',4'-Tetra-O-benzylepicatechin-4Beta→8-5,7,3',4'-tetra-O-benzylepicatechin |
| InChI Formula | InChI=1S/C8H6O3.H2O/c9-5-8(11)6-1-3-7(10)4-2-6;/h1-5,10H;1H2 |
| SMILES | O.Oc1ccc(cc1)C(=O)C=O |
| IUPAC Name | 2-(4-hydroxyphenyl)-2-oxoacetaldehyde;hydrate |
| Molecular Weight (g/mol) | 168.147 |
| Formula Weight | 168.042 |