Learn More
MedChemExpress 5(6)-TAMRA
Specifications
Specifications
| Content And Storage | -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 500 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 125 mg/mL (290.39 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 430.45 g/mol |
| CAS | 98181-63-6 |
| Synonym | 5(6)-Carboxytetramethylrhodamine |