missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%

Product Code. p-7027668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420458 1 g 1g
11430458 5 g 5g
11440458 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420458 Supplier Thermo Scientific Alfa Aesar Supplier No. B24601.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1046-56-6 | C20H14N4 | 310.36 g/mol

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1046-56-6
Molecular Formula C20H14N4
Molecular Weight (g/mol) 310.36
MDL Number MFCD00006462
InChI Key OTMYLOBWDNFTLO-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine, 1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl, 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine, as-triazine, 5,6-diphenyl-3-2-pyridyl, 5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine, maybridge1_007490, bio7c7, acmc-1boy4, cbmicro_025706
PubChem CID 70588
SMILES C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 189°C to 192°C
Quantity 1 g
Beilstein 542151
Solubility Information Soluble in water.
IUPAC Name 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine
Formula Weight 310.36
Percent Purity 99%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.