Learn More
5(6)-Carboxytetramethylrhodamine (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | 5(6)-Carboxytetramethylrhodamine |
| CAS | 98181-63-6 |
| Molecular Formula | C25H22N2O5 |
| Quantity | 500 mg |
| Synonym | 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium Inner Salt, 5(6)-TAMRA |
| SMILES | O=C(O)C(C=C1C([O-])=O)=CC=C1C2=C3C(C=C(N(C)C)C=C3)=[O+]C4=C2C=CC(N(C)C)=C4 |
| IUPAC Name | Xanthylium, 9-(2,4-dicarboxyphenyl)-3,6-bis(dimethylamino)-, inner salt (9CI, ACI) |
| Molecular Weight (g/mol) | 430.45 |
| Formula Weight | 430.15 |