Learn More
5,6,7,8,9,10-Hexahydrocyclohept[b]indole, 98%
Product Code. 11463947
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463947 1 g 1g
11473947 5 g 5g
11483947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11463947 Supplier Thermo Scientific Alfa Aesar Supplier No. A19344.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
CAS: 2047-89-4 | C13H15N | 185.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2047-89-4
Molecular Formula C13H15N
Molecular Weight (g/mol) 185.27
MDL Number MFCD00101340
InChI Key XZUJMYLNFZHNLP-UHFFFAOYSA-N
Synonym 5,6,7,8,9,10-hexahydrocyclohepta b indole, 5,6,7,8,9,10-hexahydro-cyclohepta b indole, 5h,6h,7h,8h,9h,10h-cyclohepta b indole, cycloheptan a indole, 2,3-pentano-1h-indole, 5,6,7,8,9,10-hexahydrocyclohept b indole, 5,6,7,8,9,10-hexahydrocyclohepta b indole #, 6,7,8,9,10,10a-hexahydrocyclohepta b indole, 6,7,8,9,10-pentahydrocyclohepta 1,2-b indole, 6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
PubChem CID 251955
IUPAC Name 5,6,7,8,9,10-hexahydrocyclohepta[b]indole
SMILES C1CCC2=C(CC1)NC3=CC=CC=C23
Specifications
Melting Point 143°C to 146°C
Quantity 1 g
Formula Weight 185.27
Percent Purity 98%
Chemical Name or Material 5,6,7,8,9,10-Hexahydrocyclohept[b]indole