Learn More
5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride |
| CAS | 93413-56-0 |
| Molecular Formula | C17 H25 N O2 . Cl H |
| Quantity | 5 mg |
| Synonym | (5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride, Venlafaxine Hydrochloride Imp. E (EP) as Hydrochloride |
| InChI Formula | InChI=1S/C17H25NO2.ClH/c1-18-12-16(14-6-8-15(19-2)9-7-14)17(20-13-18)10-4-3-5-11-17;/h6-9,16H,3-5,10-13H2,1-2H3;1H |
| SMILES | Cl.COc1ccc(cc1)C2CN(C)COC23CCCCC3 |
| IUPAC Name | 5-(4-methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane;hydrochloride |
| Molecular Weight (g/mol) | 311.85 |
| Formula Weight | 311.1652 |