Learn More
5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™
Product Code. 10055532
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055532 250 mg Amber Glass Bottle 250mg
10412662 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10055532 Supplier Thermo Scientific Maybridge Supplier No. MO01026CB
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Chemical Identifiers
CAS 690631-98-2
Molecular Formula C10H8N2O3
Molecular Weight (g/mol) 204.185
MDL Number MFCD05664420
InChI Key KOWRVFWZCLHEIU-UHFFFAOYSA-N
Synonym 5-3-hydroxyphenyl-1h-pyrazole-3-carboxylic acid, 5-3-hydroxy-phenyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-3-hydroxyphenyl, 3-3-hydroxyphenyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 5-3-hydroxyphenyl, 5-3-hydroxyphenyl-2h-pyrazole-3-carboxylic acid, 3-3-carboxy-1h-pyrazol-5-yl phenol, 3-carboxy-5-3-hydroxyphenyl-1h-pyrazole, 5-3-hydroxyphenyl pyrazole-3-carboxylic acid
PubChem CID 2794658
IUPAC Name 3-(3-hydroxyphenyl)-1H-pyrazole-5-carboxylic acid
SMILES C1=CC(=CC(=C1)O)C2=NNC(=C2)C(=O)O
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.18
Percent Purity 97%
Chemical Name or Material 5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid