Learn More
5-[(3-Chloro-2-fluorophenyl)methyl]-2,4-dimethoxybenzoic Acid, Mikromol
Product Code. 30461173
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461173 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461173 Supplier Mikromol Supplier No. MM3372.040025
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-[(3-Chloro-2-fluorophenyl)methyl]-2,4-dimethoxybenzoic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-[(3-Chloro-2-fluorophenyl)methyl]-2,4-dimethoxybenzoic Acid
CAS 949465-79-6
Analyte or Component Names 5-[(3-Chloro-2-fluorophenyl)methyl]-2,4-dimethoxybenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C16 H14 Cl F O4
Certifications/Compliance ISO 9001
Synonym Benzoic acid, 5-[(3-chloro-2-fluorophenyl)methyl]-2,4-dimethoxy-
InChI Formula InChI=1S/C16H14ClFO4/c1-21-13-8-14(22-2)11(16(19)20)7-10(13)6-9-4-3-5-12(17)15(9)18/h3-5,7-8H,6H2,1-2H3,(H,19,20)
Solution Type Neat
SMILES COc1cc(OC)c(cc1Cc2cccc(Cl)c2F)C(=O)O
IUPAC Name 5-[(3-chloro-2-fluorophenyl)methyl]-2,4-dimethoxybenzoic acid
Molecular Weight (g/mol) 324.7314
Quantity 25 mg
Formula Weight 324.0565 g/mol