Learn More
5-(2-Oxiranyl)-8-(phenylmethoxy)-2(1H)-quinolinone, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(2-Oxiranyl)-8-(phenylmethoxy)-2(1H)-quinolinone |
| CAS | 112281-28-4 |
| Molecular Formula | C18H15NO3 |
| Quantity | 2 mg |
| InChI Formula | InChI=1S/C18H15NO3/c20-17-9-7-14-13(16-11-22-16)6-8-15(18(14)19-17)21-10-12-4-2-1-3-5-12/h1-9,16H,10-11H2,(H,19,20) |
| SMILES | O=C1Nc2c(OCc3ccccc3)ccc(C4CO4)c2C=C1 |
| IUPAC Name | 5-(oxiran-2-yl)-8-phenylmethoxy-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 293.32 |
| Formula Weight | 293.1052 |