Learn More
5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridineethanol, TRC
Product Code. 30484268
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484268 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484268 Supplier Toronto Research Chemicals Supplier No. TRCM30364525MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridineethanol
CAS 184766-50-5
Molecular Formula C11 H15 N O3
Quantity 25 mg
Synonym 2-Pyridineethanol','5-(2-methyl-1,3-dioxolan-2-yl)-, 5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridineethanol, 2-[5-(2-Methyl-1,3-dioxolan-2-yl)pyridin-2-yl]ethan-1-ol
InChI Formula InChI=1S/C11H15NO3/c1-11(14-6-7-15-11)9-2-3-10(4-5-13)12-8-9/h2-3,8,13H,4-7H2,1H3
SMILES CC1(OCCO1)c2ccc(CCO)nc2
IUPAC Name 2-[5-(2-methyl-1,3-dioxolan-2-yl)pyridin-2-yl]ethanol
Molecular Weight (g/mol) 209.24
Formula Weight 209.1052