Learn More
DWK Life Sciences Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Product Code. p-8003877
Click to view available options
Length (Metric):
70 mm
75 mm
100 mm
130 mm
180 mm
200 mm
Diameter (Metric):
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
25 mm
30 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 11852363
Brand: DWK Life Sciences A507510000521
Only null left
Add to basket
Add to basket
Measure, mix and pour using 5.1 Borosilicate Glass Disposable Test Tubes. Tubes are rimmed and are available in various lengths.
  • Disposable
  • Clear borosilicate glass
  • Round bottom
  • Rimmed
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 75 mm
Packaging 100Pack
Rimmed Yes
Diameter (Metric) 10 mm
Product Type Test Tube