Learn More
DWK Life Sciences Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Description
- Disposable
- Clear borosilicate glass
- Round bottom
- Rimmed
Specifications
Specifications
| Design Type | Round bottom |
| Length (Metric) | 75 mm |
| Packaging | 100Pack |
| Rimmed | Yes |
| Diameter (Metric) | 10 mm |
| Product Type | Test Tube |