Learn More
(4R)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-formyloxazolidine, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | (4R)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-formyloxazolidine |
| CAS | 95715-87-0 |
| Molecular Formula | C11H19NO4 |
| Quantity | 500 mg |
| Synonym | (4R)-4-Formyl-2,2-dimethyl-3-oxazolidinecarboxylic Acid 1,1-Dimethylethyl Ester, (4R)-4-Formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylic Acid tert-Butyl Ester, (R)-Garner Aldehyde, tert-Butyl (R)-4-Formyl-2,2-dimethyloxazolidine-3-carboxylate, (+)-N-Boc-N,O-isopropylidene-D-serinal |
| InChI Formula | InChI=1S/C11H19NO4/c1-10(2,3)16-9(14)12-8(6-13)7-15-11(12,4)5/h6,8H,7H2,1-5H3/t8-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O |
| IUPAC Name | tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate |
| Molecular Weight (g/mol) | 229.27 |
| Formula Weight | 229.1314 |