Learn More
4mol/L Sodium Hydroxide, 4M, Chem Lab™
Product Code. 11927385
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity unitSize
11927385 1 L 1 L
1 options
Product Code. 11927385
Brand: Chem-Lab CL05.1424.1000
Specifications
pH >13
Color Colorless
Boiling Point 100°C
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 4M
Density 1.15g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in Water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Concentration or Composition (by Analyte or Components) 160g NaOH per 1L H2O
Normality 4 N (±0.008, 20°C)
Odor Odorless
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.