Learn More
(4aR,7S,8aR)-9,9-Dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide, 97%, BLD Pharm™
Product Code. 31032805
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032805 25 g -
31030830 10 g 10g
31022901 1 g 1g
31019290 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31032805 Supplier BLD Pharm Supplier No. BD1230925g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 104372-31-8
Molecular Formula C10H15NO3S
MDL Number MFCD30486441
Quantity 25 g
SMILES CC1(C)[C@](C2)(CC[C@@]1([H])C3)[C@]43N(O4)S2(=O)=O
Molecular Weight (g/mol) 229.30
Percent Purity 97%
Chemical Name or Material (4aR,7S,8aR)-9,9-Dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide