Learn More
Metro™ Starsys™ Supply Mobile Unit
Product Code. 15907261
Change view
Click to view available options
No. of Shelves:
4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Shelves Unit Size
15907261 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15907261 Supplier Metro™ Supplier No. SXRS76CM5
  • ONLINE EXCLUSIVE PRICE
    52680.00 NOK
    44571.92 NOK / Pack of 1
    Save 8108.08 NOK
    15% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Stand-alone products with non-locking doors.
  • 2 Wire shelves (2)
  • Non-locking doors
Specifications
Product Type Supply Mobile Unit
Color Taupe
Dimensions (L x W x H) 24.875 x 22.75 x 78.3 in.
Material Polymer
No. of Shelves 4