Learn More
Brady™ Vinyl Tapes (B-595) for BMP71 Printer - Yellow
Description
- Vinyl film with permanent adhesive
- Adheres well to pipes, walls, equipment, windows - most clean, dry surfaces
- Resistant to grease, oil and most industrial chemicals
- Five year outdoor durability in temperatures from 82° to -40°C (180° to -40°F, can be applied as low as -18°C (0°F)
Specifications
Specifications
| Color | Yellow |
| Material | Vinyl |
| Product Type | Tape |
| For Use With (Application) | Labeling pipes, walls, equipment, windows |
| For Use With (Equipment) | BMP™71 Printer |
| Quantity | 1 Ea. |
| Dimensions (LxW) | 50 ft. x 2 in. |
| Width (English) | 2 in. |
| Width (Metric) | 5.08 cm |