Learn More
BRANDTECH™ VITLAB™ Griffin Polypropylene Beakers
Product Code. p-4955129
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 12
Product Code. 15772086
Brand: BRANDTECH™ VITLAB™ 608941
Only null left
Add to basket
Add to basket
Beakers with molded graduations for durability
  • Made from high-quality polypropylene with a raised graduation scale
  • Highly transparent for excellent visibility
  • Autoclavable at 121°C
  • Complies with ISO 7056 standards
Certifications
Meets the requirements of ISO 7056
Specifications
Diameter (Metric) Base 50 mm
Capacity (Metric) 100 mL
Material Polypropylene
Height (Metric) 70 mm
Increments 20 mL
Quantity 12 Pk.
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Molded Graduations
Style Griffin